accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 66605-57-0

H3CCH3H3COONH(S)OH

CAS RN 66605-57-0

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 251.33
Molecular Formula: C14H21NO3
InChi Key: LDKDMDVMMCXTMO-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products