accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 672-47-9

OHONHNFFF

CAS RN 672-47-9

IUPAC Name: 6-hydroxy-2-(trifluoromethyl)-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-2-(trifluoromethyl)-3H-pyrimidin-4-one
Molecular Weight (g/mol): 180.09
Molecular Formula: C5H3F3N2O2
InChi Key: AMGBKPNVGVAFEN-UHFFFAOYSA-N
SMILES: OC1=CC(=O)NC(=N1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products