accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 67468-65-9

O

CAS RN 67468-65-9

IUPAC Name: 4-benzylbenzaldehyde
Synonyms: 4-benzylbenzaldehyde
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: DHARILAASAAGJJ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CC2=CC=CC=C2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products