CAS RN 705-31-7
CAS RN 705-31-7
IUPAC Name:
1-ethynyl-4-(trifluoromethyl)benzene
Synonyms:
1-ethynyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
170.13
Molecular Formula:
C9H5F3
InChi Key:
XTKBMZQCDBHHKY-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C=C1)C#C