accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 733748-92-0

CH3H3CNONHN

CAS RN 733748-92-0

IUPAC Name: 4-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]piperidine
Synonyms: 4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidine
Molecular Weight (g/mol): 195.27
Molecular Formula: C10H17N3O
InChi Key: QDRJZNBKSMAEIE-UHFFFAOYSA-N
SMILES: CC(C)C1=NOC(=N1)C1CCNCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products