accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 74-85-1

CH2H2C

CAS RN 74-85-1

IUPAC Name: ethene
Synonyms: ethylene athylen caswell no. 436 etileno epa pesticide chemical code 041901 elayl ethylene, pure bicarburretted hydrogen aethen pure ethylene
Molecular Weight (g/mol): 28.05
Molecular Formula: C2H4
InChi Key: VGGSQFUCUMXWEO-UHFFFAOYSA-N
SMILES: C=C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products