accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 769-68-6

CH3N

CAS RN 769-68-6

IUPAC Name: 2-phenylbutanenitrile
Synonyms: benzeneacetonitrile, α-ethyl-
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: IZPUPXNVRNBDSW-UHFFFAOYNA-N
SMILES: CCC(C#N)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products