CAS RN 80460-73-7
CAS RN 80460-73-7
IUPAC Name:
(4-aminophenyl)boronic acid
Synonyms:
4-aminophenylboronic acid
Molecular Weight (g/mol):
136.95
Molecular Formula:
C6H8BNO2
InChi Key:
MKPDAJWEBQRQCO-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Aminobenzeneboronic acid hydrochloride, 97%
CAS: 80460-73-7 Molecular Formula: C6H9BClNO2 Molecular Weight (g/mol): 173.403 MDL Number: MFCD03001333 InChI Key: QBYGJJSFMOVYOA-UHFFFAOYSA-N Synonym: 4-aminophenylboronic acid hydrochloride,4-aminophenyl boronic acid hydrochloride,4-aminobenzeneboronic acid hydrochloride,4-aminophenylboronic acid hcl,4-boronoaniline hydrochloride,4-aminophenylboronic acid, hcl,boronic acid, 4-aminophenyl-, hydrochloride,pubchem1747,4-aminophenylboronicacidhydrochloride,ksc494e4p PubChem CID: 2734614 IUPAC Name: (4-aminophenyl)boronic acid;hydrochloride SMILES: B(C1=CC=C(C=C1)N)(O)O.Cl