accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 850568-81-9

BCH3NHOOCH3CH3H3CHOOH

CAS RN 850568-81-9

IUPAC Name: (3-{[(tert-butoxy)carbonyl]amino}-4-methylphenyl)boronic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-4-methylphenylboronic acid
Molecular Weight (g/mol): 251.09
Molecular Formula: C12H18BNO4
InChi Key: RUPLVISWFCBMCR-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)OC(C)(C)C)C=C(C=C1)B(O)O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products