CAS RN 85553-53-3
CAS RN 85553-53-3
IUPAC Name:
3-(pyridin-2-yl)benzaldehyde
Synonyms:
3-(pyridin-2-yl)benzaldehyde
Molecular Weight (g/mol):
183.21
Molecular Formula:
C12H9NO
InChi Key:
SAPNGHSAYQXRPG-UHFFFAOYSA-N
SMILES:
O=CC1=CC=CC(=C1)C1=CC=CC=N1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 3-Pyrid-2-ylbenzaldehyde, ≥97%, Thermo Scientific™
CAS: 85553-53-3 Molecular Formula: C12H9NO Molecular Weight (g/mol): 183.21 MDL Number: MFCD02684103 InChI Key: SAPNGHSAYQXRPG-UHFFFAOYSA-N Synonym: 3-2-pyridyl benzaldehyde,3-pyridin-2-yl benzaldehyde,3-2-pyridinyl benzaldehyde,3-pyridin-2-yl-benzaldehyde,3-pyrid-2-ylbenzaldehyde,benzaldehyde, 3-2-pyridinyl,2-3-formylphenyl pyridine,acmc-209q6q,benzaldehyde,3-2-pyridinyl,5-2-pyridyl benzaldehyde PubChem CID: 3710039 IUPAC Name: 3-pyridin-2-ylbenzaldehyde SMILES: C1=CC=NC(=C1)C2=CC(=CC=C2)C=O