CAS RN 86386-77-8
CAS RN 86386-77-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole Mesilate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Thermo Scientific Alfa Aesar 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulfonate, 98%, Thermo Scientific Chemicals
CAS: 86386-77-8 MDL Number: MFCD11111978 Synonym: 1-[2-(2,4-difluorophenyl)-2-oxiranylmethyl]-1H-1,2,4-triazole methanesulfonate