accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 87199-15-3

OHBOHHO

CAS RN 87199-15-3

IUPAC Name: [3-(hydroxymethyl)phenyl]boronic acid
Synonyms: 3-(hydroxymethyl)phenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: HGTDLKXUWVKLQX-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)B(O)O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products