accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 88487-25-6

H2NOFCl

CAS RN 88487-25-6

IUPAC Name: 2-chloro-5-fluorobenzamide
Synonyms: 2-chloro-5-fluorobenzamide
Molecular Weight (g/mol): 173.57
Molecular Formula: C7H5ClFNO
InChi Key: UKUUZRYASPJJOI-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(F)=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products