CAS RN 900185-02-6
CAS RN 900185-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express PIK-294, MedChemExpress
MedChemExpress PIK-294 is a potent p110δ-selective inhibitor with an IC50 of 10 nM.
