accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 9004-99-3

H3COOOH

CAS RN 9004-99-3

IUPAC Name: 2-hydroxyethyl octadecanoate
Synonyms: glycol stearate lipal 400-S polyethylene monostearate #6000 glycol nonex 28 nonex 29 nonex 36 nonex 53 nonex 54 nonex 63 myrj solution
Molecular Weight (g/mol): 328.54
Molecular Formula: C20H40O3
InChi Key: RFVNOJDQRGSOEL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCCO
Color 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products