CAS RN 900573-88-8
CAS RN 900573-88-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express JI-101, MedChemExpress
MedChemExpress JI-101 is an orally available multi-kinase inhibitor of VEGFR2, PDGFRβ and EphB4 with potent anti-cancer activity.
