accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 91037-65-9

H2N(S)NH2NNH2ONHOHN(S)OHOONH(S)OHHOO

CAS RN 91037-65-9

IUPAC Name: (3S)-3-{2-[(2S)-2-amino-5-[(diaminomethylidene)amino]pentanamido]acetamido}-3-{[(1S)-1-carboxy-2-hydroxyethyl]carbamoyl}propanoic acid
Synonyms: (3S)-3-{2-[(2S)-2-amino-5-[(diaminomethylidene)amino]pentanamido]acetamido}-3-{[(1S)-1-carboxy-2-hydroxyethyl]carbamoyl}propanoic acid
Molecular Weight (g/mol): 433.42
Molecular Formula: C15H27N7O8
InChi Key: NNRFRJQMBSBXGO-CIUDSAMLSA-N
SMILES: N[C@@H](CCCN=C(N)N)C(=O)NCC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products