CAS RN 942947-93-5
CAS RN 942947-93-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express CCT129202, MedChemExpress
MedChemExpress CCT129202 is an aurora kinase inhibitor with IC50s of 42, 198, and 227 nM for aurora A, B and C, respectively.
