CAS RN 950736-05-7
CAS RN 950736-05-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express B-Raf IN 1, MedChemExpress
MedChemExpress B-Raf IN 1 is a potent and selective B-Raf kinase inhibitor with an IC50 of 24 nM.
