accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 958852-01-2

NOHNOSN(E/Z)

CAS RN 958852-01-2

IUPAC Name: 5-{[4-(pyridin-4-yl)quinolin-6-yl]methylidene}-1,3-thiazolidine-2,4-dione
Synonyms: 5-{[4-(pyridin-4-yl)quinolin-6-yl]methylidene}-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol): 333.37
Molecular Formula: C18H11N3O2S
InChi Key: QDITZBLZQQZVEE-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C(S1)=CC1=CC=C2N=CC=C(C3=CC=NC=C3)C2=C1
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products