CAS RN 110076-44-3
IUPAC Namn:
sodium chloro(2-methylbenzenesulfonyl)azanide
Synonymer:
sodium chloro(2-methylbenzenesulfonyl)azanide
Molekylvikt (g/mol):
227.64
Molekylär sammansättning:
C7H7ClNNaO2S
InChi Key:
BMVGZYMMLBPFDA-UHFFFAOYSA-N
SMILES:
[Na+].CC1=CC=CC=C1S(=O)(=O)[N-]Cl
1
–
3
av
3
Resultat
1
o-Chloramine T, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
o-Chloramine T, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
o-Chloramine T, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.