CAS RN 114915-16-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
N-Des-t-boc-10-deacetyl-7,10-O-bis{[(2,2,2-trichloroethyl)oxy]carbonyl} Docetaxel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Des-t-boc-10-deacetyl-7,10-O-bis{[(2,2,2-trichloroethyl)oxy]carbonyl} Docetaxel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Des-t-boc-10-deacetyl-7,10-O-bis{[(2,2,2-trichloroethyl)oxy]carbonyl} Docetaxel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.