CAS RN 1198284-77-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
tert-Butyl 4-(5-Bromo-1H-indazol-3-yl)piperidine-1-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
tert-Butyl 4-(5-Bromo-1H-indazol-3-yl)piperidine-1-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
tert-Butyl 4-(5-Bromo-1H-indazol-3-yl)piperidine-1-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.