CAS RN 120512-38-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
alpha,alpha,alpha',5-Tetramethyl-1,3-benzenediacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
alpha,alpha,alpha',5-Tetramethyl-1,3-benzenediacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
alpha,alpha,alpha',5-Tetramethyl-1,3-benzenediacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.