CAS RN 122590-77-6
IUPAC Namn:
4-azido-2,3,5,6-tetrafluorobenzoic acid
Synonymer:
4-azido-2,3,5,6-tetrafluorobenzoic acid
Molekylvikt (g/mol):
235.1
Molekylär sammansättning:
C7HF4N3O2
InChi Key:
IOJFHZXQSLNAQJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(F)=C(N=[N+]=[N-])C(F)=C1F
1
–
1
av
1
Resultat
1
4-Azido-2,3,5,6-tetrafluorobenzoic Acid(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.