CAS RN 125112-74-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
(3Alpha,5Beta,6Alpha,7Alpha)-6-Bromo-3-[(ethoxycarbonyl)oxy]-7-hydroxycholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Alpha,5Beta,6Alpha,7Alpha)-6-Bromo-3-[(ethoxycarbonyl)oxy]-7-hydroxycholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.