CAS RN 129230-50-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
1,1,2,2,3,3,4,4,5,5,5-Undecafluoro-1-pentanesulfinic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,1,2,2,3,3,4,4,5,5,5-Undecafluoro-1-pentanesulfinic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,1,2,2,3,3,4,4,5,5,5-Undecafluoro-1-pentanesulfinic Acid Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.