CAS RN 147103-31-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
4-Nitrophenyl 2-O-Alpha-D-Glucopyranosyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Nitrophenyl 2-O-Alpha-D-Glucopyranosyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Nitrophenyl 2-O-Alpha-D-Glucopyranosyl-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.