CAS RN 14919-38-1
IUPAC Namn:
[3-carboxy-2-(tetradecanoyloxy)propyl]trimethylazanium chloride
Synonymer:
[3-carboxy-2-(tetradecanoyloxy)propyl]trimethylazanium chloride
Molekylvikt (g/mol):
408.02
Molekylär sammansättning:
C21H42ClNO4
InChi Key:
TWGWHMYOGIGWDM-UHFFFAOYNA-N
SMILES:
[Cl-].CCCCCCCCCCCCCC(=O)OC(CC(O)=O)C[N+](C)(C)C
1
–
3
av
3
Resultat
1
(+/-)-Myristoylcarnitine Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+/-)-Myristoylcarnitine Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+/-)-Myristoylcarnitine Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.