CAS RN 15743-04-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-5-[2-(sulfonatooxy)ethyl]thiazolium Chloride (Thiamine Chloride Sulfate Ester), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.