CAS RN 160514-13-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2'-[(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, TRC
CAS: 160514-13-6 Molekylformel: C14 H12 N4 O Molekylvikt (g/mol): 252.27 Synonym: [2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol,Losartan Potassium Imp. B (EP) IUPAC-namn: [4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methanol LEDER: OCc1ccc(cc1)c2ccccc2c3nnn[nH]3