Läs mer
2'-[(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, TRC
Produktkod. 30452721
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452721 50 mg
30452483 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452721 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 160514-13-6
Molekylformel C14 H12 N4 O
Molekylvikt (g/mol) 252.27
Synonym [2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methanol, Losartan Potassium Imp. B (EP)
IUPAC-namn [4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methanol
LEDER OCc1ccc(cc1)c2ccccc2c3nnn[nH]3
Specifikationer
Kemiskt namn eller material (2'-(1H-Tetrazol-5-yl)biphenyl-4-yl)methanol
Kvantitet 50 mg
InChI formel InChI=1S/C14H12N4O/c19-9-10-5-7-11(8-6-10)12-3-1-2-4-13(12)14-15-17-18-16-14/h1-8,19H,9H2,(H,15,16,17,18)
Formel vikt 252.1011