CAS RN 162204-20-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
2’,3’-Di-O-acetyl-5’-deoxy-5-fluoro-N4-(pentoxycarbonxyl)cytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2’,3’-Di-O-acetyl-5’-deoxy-5-fluoro-N4-(pentoxycarbonxyl)cytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2’,3’-Di-O-acetyl-5’-deoxy-5-fluoro-N4-(pentoxycarbonxyl)cytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Pentyl [1-(2,3-Di-O-acetyl-5-deoxy-beta-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.