CAS RN 17609-52-8
IUPAC Namn:
(2R)-2-{[(benzyloxy)carbonyl]amino}-2-phenylacetic acid
Synonymer:
(R)-{[(benzyloxy)carbonyl]amino}(phenyl)acetic acid
Molekylvikt (g/mol):
285.3
Molekylär sammansättning:
C16H15NO4
InChi Key:
RLDJWBVOZVJJOS-CQSZACIVSA-N
SMILES:
OC(=O)[C@H](NC(=O)OCC1=CC=CC=C1)C1=CC=CC=C1
1
–
3
av
3
Resultat
1
Cbz-D-(-)-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cbz-D-(-)-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cbz-D-(-)-Phenylglycine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.