CAS RN 1786438-30-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Tocris Bioscience™ (R)-CR8
Cdk inhibitor; potently inhibits cdk1 and cdk2; also inhibits CK1; acts as molecular glue