CAS RN 1792999-26-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
cis-4-[[2-(1H-Benzimidazol-6-ylamino)-8-quinazolinyl]oxy]-cyclohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
cis-4-[[2-(1H-Benzimidazol-6-ylamino)-8-quinazolinyl]oxy]-cyclohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-4-[[2-(1H-Benzimidazol-6-ylamino)-8-quinazolinyl]oxy]-cyclohexanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
NCB-0846, MedChemExpress
MedChemExpress NCB-0846 is an orally available TNIK inhibitor with an IC50 of 21 nM.