CAS RN 1797102-40-9
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
4-O-tert-Butyldimethylsilyl-2,3-dihydroxy-2-methyl-butan-1,4-diol 1-O-Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-O-tert-Butyldimethylsilyl-2,3-dihydroxy-2-methyl-butan-1,4-diol 1-O-Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.