CAS RN 1797817-65-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2-[N-Benzyl-(2-hydroxyethyl)amino]-N-methyl-N-(phenylmethyl)-acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[N-Benzyl-(2-hydroxyethyl)amino]-N-methyl-N-(phenylmethyl)-acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.