CAS RN 189148-50-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
1-(2-Hydroxyethyl)-4-[3-(3-methyl-2(3H)-benzothiazolylidene)-1-propen-1-yl]quinolinium Tetrafluoroborate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.