CAS RN 191656-33-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
4-Chloroaniline-d4, TRC
CAS: 191656-33-4 Molekylformel: C6 D4 H2 Cl N Molekylvikt (g/mol): 131.5962 Synonym: Benzen-2,3,5,6-d4-amine, 4-chloro- (9CI),4-Chlorobenzen-2,3,5,6-d4-amine,4-Chloroaniline-d4,4-Chloroaniline-2,3,5,6-D4 IUPAC-namn: 4-chloro-2,3,5,6-tetradeuterioaniline LEDER: [2H]c1c([2H])c(Cl)c([2H])c([2H])c1N
2
4-Chloroaniline-2,3,5,6-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.