CAS RN 1937-62-8
IUPAC Namn:
methyl (9E)-octadec-9-enoate
Synonymer:
methyl oleate emery, oleic acid ester oleic acid ester emery Edenor MeTiO5 phytorob 926-67 (E)-methyl oleate (e)-methyl oleate edenor me 90/95V exceparl M-ol emery oleic acid ester 2301
Molekylvikt (g/mol):
296.5
Molekylär sammansättning:
C19H36O2
InChi Key:
QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES:
CCCCCCCC\C=C\CCCCCCCC(=O)OC
1
–
2
av
2
Resultat
1
Elaidic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Elaidic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.