CAS RN 202468-43-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
L-Histidine Hydrochloride Hydrate (13C6,15N3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.