CAS RN 219984-49-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Revexepride, MedChemExpress
MedChemExpress Revexepride is a highly selective 5-HT4 receptor agonist, and a potential inducer of CYP3A4 enzyme, used for the treatment of gastroesophageal reflux disease.