CAS RN 2219-31-0
IUPAC Namn:
{amino[(3-amino-3-carboxypropoxy)amino]methylidene}azanium hydrogen sulfate
Synonymer:
{amino[(3-amino-3-carboxypropoxy)amino]methylidene}azanium hydrogen sulfate
Molekylvikt (g/mol):
274.25
Molekylär sammansättning:
C5H14N4O7S
InChi Key:
MVIPJKVMOKFIEV-UHFFFAOYNA-N
SMILES:
OS([O-])(=O)=O.NC(CCONC(N)=[NH2+])C(O)=O
1
–
3
av
3
Resultat
1
L-Canavanine Sulphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Canavanine Sulphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
L-Canavanine sulfate, MedChemExpress
MedChemExpress L-Canavanine sulfate is a selective inhibitor of inducible NO synthase.