CAS RN 2227549-00-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Tocris Bioscience™ BI 01383298
Potent and selective SLC13A5 (Na+/citrate cotransporter) inhibitor
2
BI 01383298, MedChemExpress
MedChemExpress BI 01383298 is a potent inhibitor of the sodium-citrate co-transporter (SLC13A5) that is highly expressed in the liver.