CAS RN 2345-51-9
IUPAC Namn:
but-3-ynoic acid
Synonymer:
3-butynoic acid but-3-insaeure
Molekylvikt (g/mol):
84.07
Molekylär sammansättning:
C4H4O2
InChi Key:
KKAHGSQLSTUDAV-UHFFFAOYSA-N
SMILES:
OC(=O)CC#C
1
–
3
av
3
Resultat
1
3-Butynoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Butynoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Butynoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.