CAS RN 23576-23-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
4-Chloro-5-(dimethylamino)-2-(3-(trifluoromethyl)phenyl)pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Chloro-5-(dimethylamino)-2-(3-(trifluoromethyl)phenyl)pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Chloro-5-(dimethylamino)-2-(3-(trifluoromethyl)phenyl)pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.