CAS RN 2370-63-0
IUPAC Namn:
2-ethoxyethyl 2-methylprop-2-enoate
Synonymer:
ethoxyethyl methacrylate
Molekylvikt (g/mol):
158.2
Molekylär sammansättning:
C8H14O3
InChi Key:
SFPNZPQIIAJXGL-UHFFFAOYSA-N
SMILES:
CCOCCOC(=O)C(C)=C
1
–
3
av
3
Resultat
1
2-Ethoxyethyl Methacrylate (stabilized with MEHQ), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Ethoxyethyl Methacrylate (stabilized with MEHQ), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Ethoxyethyl Methacrylate (stabilized with MEHQ), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.