CAS RN 23749-65-7
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2,4,6-Trichlorotoluene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
1,3,5-Trichloro-2-methylbenzene, TRC
CAS: 23749-65-7 Molekylformel: C7 H5 Cl3 Molekylvikt (g/mol): 195.47 Synonym: 1,3,5-Trichloro-2-methylbenzene (ACI),Toluene, 2,4,6-trichloro- (8CI),2,4,6-Trichlorotoluene,1,3,5-Trichloro-2-methylbenzene (ACI),Toluene, 2,4,6-trichloro- (8CI),2,4,6-Trichlorotoluene,2,4,6-Trichloro-toluene; IUPAC-namn: 1,3,5-trichloro-2-methylbenzene LEDER: Cc1c(Cl)cc(Cl)cc1Cl