CAS RN 2410284-89-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
2,2'-(Piperazine-1,4-diyl)bis(N-(4-aminophenyl)-N-methylacetamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2'-(Piperazine-1,4-diyl)bis(N-(4-aminophenyl)-N-methylacetamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,2'-(Piperazine-1,4-diyl)bis(N-(4-aminophenyl)-N-methylacetamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.